Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Empty solid phase extraction cartridges, Bond Elut™

Cartridge, 12 ml
Catalog # 551-0359
Supplier:   VARIAN
Empty solid phase extraction cartridges, Bond Elut™
Catalog # 551-0359
Supplier:  VARIAN
Empty solid phase extraction cartridges, Bond Elut™
Empty solid phase extraction cartridges, Bond Elut™

Cartridge, 12 ml

Price
148,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

Bond Elut empty SPE cartridges are made with high purity polypropylene for cleaner extracts.

  • Uniform batch-to-batch size for consistent performance
  • Cartridges are available from 1 to 60 ml

Ordering information: Frits has to be ordered separately

Specifications
  • Column Format:
    Cartridge
  • Capacity:
    12 ml
  • Seq:
    04
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Adapter caps for 1, 3 and 6 ml Bond Elut™ cartridges
    Adapter caps for 1, 3 and 6 ml Bond Elut™ cartridges
    551-0354
    Log in for availability
    1 * 15 ST - 98,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Luer stopcocks
    Luer stopcocks
    551-0355
    Log in for availability
    1 * 15 ST - 142,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Adapter caps for3 ml Gilson ASPEC SPE systems
    Adapter caps for3 ml Gilson ASPEC SPE systems
    VARI12131035
    Log in for availability
    1 * 1.000 ST - 804,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Adapter caps for1ml Gilson ASPEC SPE systems
    Adapter caps for1ml Gilson ASPEC SPE systems
    VARI12131034
    Log in for availability
    1 * 1.000 ST - 739,00 €

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.