Empty solid phase extraction cartridges, Bond Elut™
Cartridge, 6 ml
Catalog # 551-0358
Supplier: VARIAN
Jump to:
Product Details & Documents
Bond Elut empty SPE cartridges are made with high purity polypropylene for cleaner extracts.
- Uniform batch-to-batch size for consistent performance
- Cartridges are available from 1 to 60 ml
Ordering information: Frits has to be ordered separately
Specifications
- Column Format:Cartridge
- Capacity:6 ml
- Seq:03
Accessories
Product Information
- Catalog #AvailabilityPrice
- Adapter caps for 1, 3 and 6 ml Bond Elut™ cartridgesAdapter caps for 1, 3 and 6 ml Bond Elut™ cartridges551-0354Log in for availability1 * 15 ST - 98,00 €
Product Information
- Catalog #AvailabilityPrice
- Luer stopcocks
Product Information
- Catalog #AvailabilityPrice
- Adapter caps for3 ml Gilson ASPEC SPE systemsAdapter caps for3 ml Gilson ASPEC SPE systemsVARI12131035Log in for availability1 * 1.000 ST - 804,00 €
Product Information
- Catalog #AvailabilityPrice
- Adapter caps for1ml Gilson ASPEC SPE systemsAdapter caps for1ml Gilson ASPEC SPE systemsVARI12131034Log in for availability1 * 1.000 ST - 739,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



