Solid phase extraction cartridges, Bond Elut™ Florisil
Straight barrel cartridge, Florisil, 200 µm, 1 ml, 100 mg
Catalog # VARI12102024
Supplier: VARIAN
Jump to:
Product Details & Documents
Florisil is a magnesia-loaded silica gel, extremely polar in nature and ideal for the isolation of polar compounds from non-polar matrices.
- Pesticide residue (PR) grade
- Fast flow, ideal for viscous samples
The larger particle size of the sorbent enables flow for large sample volumes and is therefore an attractive alternative to silica if the sample matrix is particularly viscous.
Specifications
- Particle Size:200 µm
- Capacity:1 ml
- Description:Straight barrel cartridge
- Seq:02
- Bed weight:100 mg
- Phase:Florisil
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



