Orders LinkContactUsLinkComponent
Test kits, digital titration method
Test kits, digital titration method
Total hardness HA-DT: EDTA method, 10 - 4000 mg/l, 1 KIT
Catalog # HACH2063600
Supplier:   Hach
Test kits, digital titration method
Catalog # HACH2063600
Supplier:  Hach
CAS Number:  
Test kits, digital titration method
Test kits, digital titration method
Total hardness HA-DT: EDTA method, 10 - 4000 mg/l, 1 KIT
CAS Number:  
Price
631,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Total hardness HA-DT: EDTA method
  • Range:
    10 - 4000 mg/l