Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer powder pillows

Buffer powder pillows

10,01, ±0,02, Carbonate, Blue, 25 °C, 15
Catalog # HACH2227195
Supplier:   Hach
Buffer powder pillows
Catalog # HACH2227195
Supplier:  Hach
CAS Number:  
Buffer powder pillows
Buffer powder pillows

10,01, ±0,02, Carbonate, Blue, 25 °C, 15

CAS Number:  
Price
25,50 €
Log in for availability
1 * 15 ST
Product Details & Documents

The unit-dose powder pillows assure buffer freshness and eliminate the possibility of contamination. Each powder pillow prepares 50 ml of buffer solutions of respective pH.

  • Fresh and reliable
  • No danger of contamination
  • Traceable to NIST standards

Delivery information: The buffer pillows are delivered in an airtight container and are supplied with a certificate of conformity.

Specifications

Specifications

  • pH buffer type:
    Carbonate
  • Pk:
    15
  • Reference temperature:
    25 °C
  • Colour:
    Blue
  • pH accuracy:
    ±0,02
  • pH value:
    10,01

Specifications