Water hardness testing solutions
Hardness 3 solution (EDTA), 1 l, EDTA, 1 L
N° de catalogue HACH42653
Fournisseur: Hach
Aller à :
Product Details & Documents
Spécifications
Spécifications
- Chemical method:EDTA
- Pk:1 L
- Description:Hardness 3 solution (EDTA), 1 l
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



