Order Entry
Belgium
Orders LinkContactUsLinkComponent
Test kits, drop count tests

Test kits, drop count tests

Hardness 3 solution, 100 ml MDB, 100 mL
Catalog # HACH42632
Supplier:   Hach
Test kits, drop count tests
Catalog # HACH42632
Supplier:  Hach
CAS Number:  
Test kits, drop count tests
Test kits, drop count tests

Hardness 3 solution, 100 ml MDB, 100 mL

CAS Number:  
Price
26,30 €
Log in for availability
1 * 100 mL
Product Details & Documents

The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.

  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit

Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Hardness 3 solution, 100 ml MDB

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.