Order Entry
Belgium
Orders LinkContactUsLinkComponent
Test kits, digital titration method

Test kits, digital titration method

Sulfuric acid digital titrator cartridge, 0,1600 N, 13 mL
Catalog # HACH1438801
Supplier:   Hach
Test kits, digital titration method
Catalog # HACH1438801
Supplier:  Hach
CAS Number:  
Test kits, digital titration method
Test kits, digital titration method

Sulfuric acid digital titrator cartridge, 0,1600 N, 13 mL

CAS Number:  
Price
40,50 €
Log in for availability
1 * 13 mL
Product Details & Documents

Digital titration provides a more precise and accurate method than the drop test method.

  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field

Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.

Specifications

Specifications

  • Pk:
    13 mL
  • Description:
    Sulfuric acid digital titrator cartridge, 0,1600 N

Specifications