Order Entry
Belgium
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ PBA

Solid phase extraction cartridges, Bond Elut™ PBA

LRC cartridge, PBA, 60 Å, 40 µm, 10 ml, 100 mg
Catalog # 551-0344
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ PBA
Catalog # 551-0344
Supplier:  VARIAN
Solid phase extraction cartridges, Bond Elut™ PBA

LRC cartridge, PBA, 60 Å, 40 µm, 10 ml, 100 mg

Price
345,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

PBA is a unique sorbent consisting of a phenylboronic acid covalently linked to a silica gel surface. Covalent processes result in a higher retention to the sorbent surface than other mechanisms, resulting in superior clean-up. The boronate group has a high specificity for cis-diol containing compounds such as catechols, nucleic acids, low molecular weight proteins and carbohydrates.

  • High specificity for cis-diol compounds
  • Very effective in separating DNA and RNA

Bond Elut™ PBA can also be used to retain oligomeric materials such as polyethylene glycols (PEGs) from a variety of aqueous or biological matrices.

Specifications
  • Particle Size:
    40 µm
  • Pore size:
    60 Å
  • Capacity:
    10 ml
  • Description:
    LRC cartridge
  • Seq:
    01
  • Bed weight:
    100 mg
  • Phase:
    PBA
  • Carbon load:
    7,9%
  • Cat. No.:
    551-0344
  • Endcapped:
    No
  • Surface area:
    500 m²/g
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.