Micro pipettes, disposable, Microcaps®
6,66 µl, 72 mm
N° de catalogue DRUM1-000-0066
Fournisseur: DRUMMOND
Aller à :
Product Details & Documents
Micro pipettes are precision-bore glass capillary tubes cut to predetermined lengths so that fill volumes are constant and precise. Capillary action draws the fluid into the tube and a squeeze of the bulb dispenses it with accuracy of ±1%.
For fast, safe, clean transfer of liquids in micro quantities. Suitable for both general and hazardous microchemical applications.
Ordering information: Delivery includes 100 pipettes and one bulb dispenser.
Spécifications
- Capacity:6,66 µl
- Length:72 mm
- Seq:15
Accessories
Informations sur le produit
- N° de catalogueDisponibilitéPrix
- Silicone tips for Microcaps®



