Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Micro pipettes, disposable, Microcaps®

3,33 µl, 36 mm
Catalog # DRUM1-000-0033
Supplier:   DRUMMOND
Micro pipettes, disposable, Microcaps®
Catalog # DRUM1-000-0033
Supplier:  DRUMMOND
Micro pipettes, disposable, Microcaps®
Micro pipettes, disposable, Microcaps®

3,33 µl, 36 mm

Price
31,10 €
Log in for availability
1 * 100 ST
Product Details & Documents

Micro pipettes are precision-bore glass capillary tubes cut to predetermined lengths so that fill volumes are constant and precise. Capillary action draws the fluid into the tube and a squeeze of the bulb dispenses it with accuracy of ±1%.

For fast, safe, clean transfer of liquids in micro quantities. Suitable for both general and hazardous microchemical applications.

 : Delivery includes 100 pipettes and one bulb dispenser.

Specifications
  • Capacity:
    3,33 µl
  • Length:
    36 mm
  • Seq:
    10
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Silicone tips for Microcaps®
    Silicone tips for Microcaps®
    DRUM1-000-9003
    Log in for availability
    1 * 3 ST - 6,00 €