Microtest manifolds
Microtest manifold, SS/PTFE, straight, 16-place for 384 well plate, 4,5 mm centre-to-centre, pin length 11 mm
Catalog # DRUM3-000-102
Supplier: DRUMMOND
Aller à :
Product Details & Documents
Stainless steel/PTFE manifolds allow multiple test positions to be fed, rinsed, or aspirated quickly and easily.
- Autoclavable
- Use for both sample delivery and aspiration operations
- Supplied complete with cleaning wire for easy cleaning
- Positive Luer lock connection
Spécifications
- Description:Microtest manifold, SS/PTFE, straight, 16-place for 384 well plate, 4,5 mm centre-to-centre, pin length 11 mm
- Seq:05