Orders LinkContactUsLinkComponent
Microtest manifolds
Microtest manifolds
Microtest manifold, SS/PTFE, bent, 8-place for 96 well plate, 9 mm centre-to-centre, pin length 11 mm
Catalog # DRUM3-000-094
Supplier:   DRUMMOND
Microtest manifolds
Catalog # DRUM3-000-094
Supplier:  DRUMMOND
Microtest manifolds
Microtest manifold, SS/PTFE, bent, 8-place for 96 well plate, 9 mm centre-to-centre, pin length 11 mm
Price
529,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel/PTFE manifolds allow multiple test positions to be fed, rinsed, or aspirated quickly and easily.
  • Autoclavable
  • Use for both sample delivery and aspiration operations
  • Supplied complete with cleaning wire for easy cleaning
  • Positive Luer lock connection
Specifications
  • Description:
    Microtest manifold, SS/PTFE, bent, 8-place for 96 well plate, 9 mm centre-to-centre, pin length 11 mm
  • Seq:
    08