Test kits, drop count tests
Chloride, model 8-P, use silver nitrate titrant, 5 - 100 mg/l Cl-
20 - 400 mg/l Cl-, 1 KIT
20 - 400 mg/l Cl-, 1 KIT
Catalog # HACH144001
Supplier: Hach
Jump to:
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
- Portable - ideal for use in field or in lab
- Used as a quick reference or for confirmatory test with colour indicator
- Reagents supplied with the kit
: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
- Pk:1 KIT
- Description:Chloride, model 8-P, use silver nitrate titrant
- Range:5 - 100 mg/l Cl-<br />20 - 400 mg/l Cl-