Order Entry
Belgium
Orders LinkContactUsLinkComponent
Micro pipettes, disposable, Microcaps®

Micro pipettes, disposable, Microcaps®

8,0 µl, 44 mm
Catalog # DRUM1-000-0080
Supplier:   DRUMMOND
Micro pipettes, disposable, Microcaps®
Catalog # DRUM1-000-0080
Supplier:  DRUMMOND
Micro pipettes, disposable, Microcaps®
Micro pipettes, disposable, Microcaps®

8,0 µl, 44 mm

Price
Inquire For Price
Log in for availability
1 * 100 ST
Product Details & Documents

Micro pipettes are precision-bore glass capillary tubes cut to predetermined lengths so that fill volumes are constant and precise. Capillary action draws the fluid into the tube and a squeeze of the bulb dispenses it with accuracy of ±1%.

For fast, safe, clean transfer of liquids in micro quantities. Suitable for both general and hazardous microchemical applications.

Ordering information: Delivery includes 100 pipettes and one bulb dispenser.

Specifications
  • Capacity:
    8,0 µl
  • Length:
    44 mm
  • Seq:
    17
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Silicone tips for Microcaps®
    Silicone tips for Microcaps®
    DRUM1-000-9003
    Log in for availability
    1 * 3 ST - 6,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.