Paraformaldehyde, powder, TECHNICAL
Plastic bucket, 5 kg
Supplier: VWR Chemicals
Paraformaldehyde, powder, TECHNICAL
Catalog # 28794.364
Supplier: VWR Chemicals
CAS Number: 30525-89-4
Jump to:
Product Details & Documents
Synonyms: Polyoxymethylene
- CAS Number:30525-89-4
- Formula:(CH₂O)n
- MDL Number:MFCD00133991
- Merck Index:13,07096
- Boiling Pt:135…136 °C (13 Torr)
- Melting Pt:120…170 °C
- Density:0.88 g/cm³ (25 °C)
- Flash Pt:70 °C (closed cup)
- Storage Temperature:Ambient
- UN:2213
- ADR:4.1,III
Specifications
Specification Test Results
- Identification:Passes test
Specifications
- Pack type:Plastic bucket
- Pk:5 kg



