Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium hydroxide 35% in aqueous solution, TECHNICAL

Sodium hydroxide 35% in aqueous solution, TECHNICAL

Plastic drum, 200 L
Catalog # 28221.558
Corrosive
Supplier:   VWR Chemicals
Sodium hydroxide 35% in aqueous solution, TECHNICAL
Catalog # 28221.558
Supplier:  VWR Chemicals
CAS Number:  1310-73-2
Sodium hydroxide 35% in aqueous solution, TECHNICAL
Sodium hydroxide 35% in aqueous solution, TECHNICAL

Plastic drum, 200 L

CAS Number:  1310-73-2
Price
Inquire For Price
Log in for availability
1 * 200 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:119…135 °C (1013 hPa)
  • Melting Pt:9…10 °C
  • Density:1,53 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II
Specifications

Specification Test Results

  • Assay:
    Min. 34 %
  • Heavy metals (as Pb):
    Max. 50 ppm
  • Fe (Iron):
    Max. 50 ppm

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    200 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.