Colour coders for cryovials, CryoColorCode, Nunc™
Violet
Catalog # 479-2487
Supplier: Thermo Fisher Scientific
Colour coders for cryovials, CryoColorCode, Nunc™
Catalog # 479-2487
Supplier: Thermo Fisher Scientific
Aller à :
Product Details & Documents
Colour coded inserts for the cap with inscription area.
Notch to remove tube or CryoColorCodes.
Documents
Spécifications
- Colour:Violet
- Seq:15
- Cat. No.:479-2487
- Environmental certification:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Environmentally Preferable:Low Manufacturing Impact