Order Entry
Belgium
Orders LinkContactUsLinkComponent
Graphite, powder, TECHNICAL

Graphite, powder, TECHNICAL

Plastic bag, 5 kg
Catalog # 24489.363
Supplier:   VWR Chemicals
Graphite, powder, TECHNICAL
Catalog # 24489.363
Supplier:  VWR Chemicals
CAS Number:  7782-42-5
Graphite, powder, TECHNICAL
Graphite, powder, TECHNICAL

Plastic bag, 5 kg

CAS Number:  7782-42-5
Price
234,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
  • CAS Number:7782-42-5
  • Formula:C
  • MDL Number:MFCD00144065
  • Molecular Weight:12.01 g/mol
  • Merck Index:12,04560
  • Melting Pt:3827 °C
  • Density:2.09…2.23 g/cm³ (20 °C)
  • EINECS:231-955-3
Specifications

Specification Test Results

  • Identification:
    Passes test

Specifications

  • Pack type:
    Plastic bag
  • Pk:
    5 kg

Specifications