Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample cuvettes

Sample cuvettes

Sample cuvette, with lid, 35 ml, 100 mm, Glass
Catalog # HACH2629201
Supplier:   Hach
Sample cuvettes
Catalog # HACH2629201
Supplier:  Hach
Sample cuvettes
Sample cuvettes

Sample cuvette, with lid, 35 ml, 100 mm, Glass

Price
527,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Sample cuvettes for photometric measurements.

  • Available with or without lid
  • Available reusable (made from glass) or for single use (made from plastics)
Specifications
  • Path length:
    100 mm
  • Volume:
    35 ml
  • Window material:
    Glass
  • Description:
    Sample cuvette, with lid
  • Seq:
    23
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.