Orders LinkContactUsLinkComponent
Racks for cooling circulators
Racks for cooling circulators
Stainless steel rack for 1×in AL 5, 3×in AL 12, 5×in AL 18, AL 25, 10, 24/30 mm, 110 mm high
Catalog # 462-1011
Supplier:   LAUDA
Racks for cooling circulators
Catalog # 462-1011
Supplier:  LAUDA
Racks for cooling circulators
Stainless steel rack for 1×in AL 5, 3×in AL 12, 5×in AL 18, AL 25, 10, 24/30 mm, 110 mm high
Price
308,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • For tube Ø:
    24/30 mm
  • For tubes:
    110 mm high
  • No. of Holes:
    10
  • Description:
    Stainless steel rack for 1×in AL 5, 3×in AL 12, 5×in AL 18, AL 25
  • Seq:
    04