Orders LinkContactUsLinkComponent
Racks for cooling circulators
Racks for cooling circulators
Stainless steel rack for 1×in AL 5, 3×in AL 12, 5×in AL 18, AL 25, 21, 14/18 mm, 80 mm high
N° de catalogue 462-1009
Fournisseur:   LAUDA
Racks for cooling circulators
N° de catalogue 462-1009
Fournisseur:  LAUDA
Racks for cooling circulators
Stainless steel rack for 1×in AL 5, 3×in AL 12, 5×in AL 18, AL 25, 21, 14/18 mm, 80 mm high
Prix
308,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • For tube Ø:
    14/18 mm
  • For tubes:
    80 mm high
  • No. of Holes:
    21
  • Description:
    Stainless steel rack for 1×in AL 5, 3×in AL 12, 5×in AL 18, AL 25
  • Seq:
    02