Orders LinkContactUsLinkComponent
Electrode Adapter Cables
Electrode Adapter Cables
Adapter, BNC meter input + screw cap electrode connector; detachable cable to BNC connector
Catalog # 662-1535
Supplier:   Thermo Orion
Electrode Adapter Cables
Catalog # 662-1535
Supplier:  Thermo Orion
Electrode Adapter Cables
Adapter, BNC meter input + screw cap electrode connector; detachable cable to BNC connector
Price
112,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These cables can be used to convert an electrode with one connector to the required connector of another meter.
For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2 mm reference pins or just a stripped lead inputs.
Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors.
Specifications
  • Description:
    Adapter, BNC meter input + screw cap electrode connector; detachable cable to BNC connector
  • Seq:
    05