Orders LinkContactUsLinkComponent
Watch glasses
Watch glasses
Watch glass, PYREX® borosilicate glass with firepolished edges, 100 mm
Catalog # 216-2751
Supplier:   SciLabware
Watch glasses
Catalog # 216-2751
Supplier:  SciLabware
Watch glasses
Watch glasses
Watch glass, PYREX® borosilicate glass with firepolished edges, 100 mm
Price
159,00 €
Log in for availability
1 * 10 ST
Product Details & Documents
PYREX®, borosilicate glass, with fused edges.
  • Excellent chemical resistance
  • High temperature resistance
For evaporation work and covering heated beakers. Fire-polished edges to reduce chipping and breakage.
Specifications
  • Ø:
    100 mm
  • Description:
    Watch glass, PYREX® borosilicate glass with firepolished edges
  • Seq:
    07