Hazardous spill kits
PF2001 folded chemical sorbent, multi-format, 119 litres/kit
Catalog # 121-1048
Supplier: 3M
Jump to:
Product Details & Documents
These absorption kits can be deployed wherever hazardous liquids are in use and can be used to contain and clean up a spill without delay.
- P110: For smaller spills or final clear-up of larger ones, sheets of 280×330 mm, absorption of 0,25 litre/sheet
- P130: Roll for large areas, can be cut to size, 33×3000 cm, absorption 50 litre/roll
- P190: Universal roll, 48×3300 cm, absorption 72 litre/roll
- P200: Minibooms, 75×1200 mm (Ø×l), absorption of 3,75 litre/miniboom
- P300: Pillows of 180×380 mm, absorption of 2 litre/pillow
- P-F2001: Folded sorbent that can be used as mini-booms, pillows, sheets or rolls, 12,5×1520 cm, perforated every 40,6 cm, absorption 119 litres/pack
Documents
Specifications
- Description:PF2001 folded chemical sorbent, multi-format
- Seq:08
- Absorbency:119 litres/kit
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



