Spoons, POLY
Straight, 185 mm, 40×20 mm
231-1069
: BOCHEM
Stainless steel 18/10, anti-magnetic. Multipurpose spoon for a wide range of uses. Special shape for light and easy handling.
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Quick and easy to clean
- Description:Straight
- Length:185 mm
- Seq:02
- Spoon size:40×20 mm



