Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Roller bottles, WHEATON®

Roller bottles, with 51 mm, shallow skirted, rubber lined, black phenolic cap, 4230 ml, 1555 cm², 110×535 mm, 4/case
  734-3025
Roller bottles, WHEATON®
  734-3025
Roller bottles, WHEATON®
Roller bottles, WHEATON®

Roller bottles, with 51 mm, shallow skirted, rubber lined, black phenolic cap, 4230 ml, 1555 cm², 110×535 mm, 4/case

 
798,00 €
   
1 * 4 ST
 

Rollers bottles are specifically designed for adherent cell roller culture

  • Bottles withstands repeated wet or dry sterilization cycles and are reusuable
  • These culture vessels are made to exacting tolerances in order to provide higher cell yields
  • Made of USP Type I borosilicate glass

 : *The product(s) on this page are outside the scope of the EU Directive 2019/904

 
  • Growth area:
    1555 cm²
  • Packed:
    4/case
  • Capacity:
    4230 ml
  • Description:
    Roller bottles, with 51 mm, shallow skirted, rubber lined, black phenolic cap
  • Ø ext.×H:
    110×535 mm