Orders LinkContactUsLinkComponent
Charts
Charts
C181, –30...+50 °C, 7 days
Catalog # 620-0390
Supplier:   DICKSON
Charts
Catalog # 620-0390
Supplier:  DICKSON
Charts
C181, –30...+50 °C, 7 days
Price
73,40 €
Log in for availability
1 * 60 ST
Product Details & Documents
Specifications
  • Description:
    C181
  • Seq:
    07
  • Range:
    –30...+50 °C
  • Recording time:
    7 days