Multiscreen® Filter Plates, Opaque
Glass fiber filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated or particular laden samples. They are also widely used for clarification of aqueous solutions.
- Multiple configurations and applications
- Chemical compatibility
- Automation-compatible
- Superior vacuum filtration and filtrate collection
- Hydrophilic lid
- Colour code: Opaque
Glass fiber filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated or particulate-laden samples. They are also widely used for clarification of aqueous solutions.
Application:
Receptor/ligand binding assays, sample preparation, TCA precipitation of protein, DNA purification.
Research category and general filtration.
Nucleic acid and assays.
Binding type: High binding surface
Height: 14,4 mm
Length: 128 mm
Width: 85,5 mm
- Application:Receptor/Ligand Binding Assays, sample Preparation, TCA precipitation of protein
- Membrane:PVDF
- No. of wells:96
- Pore size:―
- Sterile:-
- Colour:Opaque
- Description:GFC plate, filtration area 0,26 cm²
- Material:Titanium
- Seq:02
- Well volume:50 - 250 μl
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



