Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Bottles, narrow neck, with thread

125 ml
Catalog # 215-2609
Supplier:   GRAVIS
Bottles, narrow neck, with thread
Catalog # 215-2609
Supplier:  GRAVIS
Bottles, narrow neck, with thread
Bottles, narrow neck, with thread

125 ml

Price
118,00 €
Log in for availability
1 * 104 ST
Product Details & Documents

Glass, clear, amber or yellow.

  • Neck Ø: 28 mm
  • Good chemical and temperature resistance
  • Amber coloured to protect light sensitive substances

Choice of various screw caps.

 : Supplied without screw caps - please order separately.

 : Packaged in impermeable plastic packs.

Specifications
  • Capacity:
    125 ml
  • Seq:
    05