Roller bottles, WHEATON®
Roller bottles, with 38 mm deep skirted, rubber lined black phenolic cap, 1380 ml, 550 cm², 110×240 mm, 4/case
734-3017
Rollers bottles are specifically designed for adherent cell roller culture
- Bottles withstands repeated wet or dry sterilization cycles and are reusuable
- These culture vessels are made to exacting tolerances in order to provide higher cell yields
- Made of USP Type I borosilicate glass
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
- Growth area:550 cm²
- Packed:4/case
- Capacity:1380 ml
- Description:Roller bottles, with 38 mm deep skirted, rubber lined black phenolic cap
- Ø ext.×H:110×240 mm
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



