Orders LinkContactUsLinkComponent
Mini molecular models: DNA
Mini molecular models: DNA
Mini molecular model of DNA, 22 layers
Catalog # 763-0454
Marketsource
Supplier:   SPIRING ENTERPRISES
Mini molecular models: DNA
Catalog # 763-0454
Marketsource
Mini molecular models: DNA
Mini molecular models: DNA
Mini molecular model of DNA, 22 layers
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Mini molecular models of DNA with nucleic acids, five-sided sugar molecules and pyramid-shaped phosphate molecules.
Only correct base pairs can be connected.
Ordering information: Components include colour-coded bases: G, C, A, T.
Specifications
  • Description:
    Mini molecular model of DNA, 22 layers