Orders LinkContactUsLinkComponent
Staining racks
Staining racks
Tray, PVC, 380×170×80 mm
Catalog # 631-0337
Staining racks
Catalog # 631-0337
Staining racks
Staining racks
Tray, PVC, 380×170×80 mm
Price
42,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Staining rack, Plexiglas®, in the form of an extendable bridge, that can be fixed in the horizontal or at an inclined position. Suitable for trays or dishes of 380 to 620 mm in length.
Specifications
  • Description:
    Tray, PVC
  • W×D×H:
    380×170×80 mm