Orders LinkContactUsLinkComponent
Comparator kits, Lovibond® system 2000
Comparator kits, Lovibond® system 2000
European Pharmacopoeia (EP) Colour Comparator 2000+ with Daylight 2000 Lighting unit; EP discs: 4/64A, R1-R7; 4/64B, Y1-Y7; 4/64C, B1-B9; 4/64D, BY1-BY7; 4/64D, GY1-GY7; 2 x W680/OG/40 mm fused cells, with 40 mm path length, AF332
Catalog # 633-0023
Supplier:   Lovibond Tintometer
Comparator kits, Lovibond® system 2000
Catalog # 633-0023
Comparator kits, Lovibond® system 2000
Comparator kits, Lovibond® system 2000
European Pharmacopoeia (EP) Colour Comparator 2000+ with Daylight 2000 Lighting unit; EP discs: 4/64A, R1-R7; 4/64B, Y1-Y7; 4/64C, B1-B9; 4/64D, BY1-BY7; 4/64D, GY1-GY7; 2 x W680/OG/40 mm fused cells, with 40 mm path length, AF332
Price
3 550,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Available for many of the commonly used colour scales, these kits are a convenient means of ordering the complete range of equipment required. Each system includes.
  • A visual comparator instrument which brings the sample and colour standards into the same field of view for more accurate colour matching
  • A selection of colour standards - stable-coloured glass filters in test discs, which have been pre-calibrated to give direct readings over set measuring ranges
  • A standardised benchtop light source to ensure constant, uniform lighting conditions for colour matching and, therefore, consistent and accurate readings
  • Glassware of the appropriate type and path length
Specifications
  • Description:
    European Pharmacopoeia (EP) Colour Comparator 2000+ with Daylight 2000 Lighting unit; EP discs: 4/64A, R1-R7; 4/64B, Y1-Y7; 4/64C, B1-B9; 4/64D, BY1-BY7; 4/64D, GY1-GY7; 2 x W680/OG/40 mm fused cells, with 40 mm path length
  • Type:
    AF332