Orders LinkContactUsLinkComponent
Drying tubes, U-shaped
Drying tubes, U-shaped
180, 18 mm
  LENZ05339018
Drying tubes, U-shaped
  LENZ05339018
Drying tubes, U-shaped
Drying tubes, U-shaped
180, 18 mm
29,50 €
1 * 1 ST
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Drying tubes, U-shaped, side tubes with olives (Ext.Ø: 6 mm).
  • Ø int.:
    18 mm
  • Length:
    180