Volumetric overflow flasks, without stopper
365 ml, Overflow flask
NEUB0372-10-365
These volumetric overflow flasks are made of borosilicate glass 3.3.
- Calibrated to deliver (TD, Ex)
- Cut and ground exactly at the volume-mark
- Without stopper
Specially designed for water analysis.
- Capacity:365 ml
- Description:Overflow flask
- Seq:06