Orders LinkContactUsLinkComponent
Sample spoons
Sample spoons
Sample spoon, 180 mm
Catalog # BURK5320-0180
Supplier:   BURKLE
Sample spoons
Catalog # BURK5320-0180
Supplier:  BURKLE
Sample spoons
Sample spoons
Sample spoon, 180 mm
Price
10,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Made of AISI 304 (1.4301) stainless steel. This specially shaped spoon reaches almost every corner in containers and prevents samples from trickling out.
  • For withdrawing small sample quantities
  • Largely chemical resistant (not against acids and alkalis)
Specifications
  • Description:
    Sample spoon
  • Length:
    180 mm
  • Seq:
    2