Orders LinkContactUsLinkComponent
Accessories for analytical Mill, A 11 basic
Accessories for analytical Mill, A 11 basic
Funnel, A 11.7, Use with liquid nitrogen, prevents splashing when pouring
Catalog # 431-2114
Supplier:   IKA
Accessories for analytical Mill, A 11 basic
Catalog # 431-2114
Supplier:  IKA
Accessories for analytical Mill, A 11 basic
Accessories for analytical Mill, A 11 basic
Funnel, A 11.7, Use with liquid nitrogen, prevents splashing when pouring
Price
301,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Funnel, A 11.7
  • Seq:
    07
  • For:
    Use with liquid nitrogen, prevents splashing when pouring
  • Cat. No.:
    431-2114