Order Entry
Belgium
Orders LinkContactUsLinkComponent
Rods, AR-Glas®

Rods, AR-Glas®

8 mm, 500 mm
  441-0272
Rods, AR-Glas®
  441-0272
Rods, AR-Glas®

8 mm, 500 mm

 
 
   
1 * 10 ST
 

AR glass.

Lenght: 500 mm

 
  • Ext.Ø:
    8 mm
  • Length:
    500 mm
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.