Order Entry
Belgium
Orders LinkContactUsLinkComponent
Rods, AR-Glas®

Rods, AR-Glas®

7 mm, 500 mm
Catalog # 441-0271
Supplier:   EMMANUEL PIERRE
Rods, AR-Glas®
Catalog # 441-0271
Supplier:  EMMANUEL PIERRE
Rods, AR-Glas®

7 mm, 500 mm

Price
Inquire For Price
Log in for availability
1 * 10 ST
Product Details & Documents

AR glass.

Lenght: 500 mm

Specifications
  • Ext.Ø:
    7 mm
  • Length:
    500 mm
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.