ContactUsLinkComponent
PH/ION METER 340I,ROBUST AND WATERPROOF PH/ION HANDHELD INSTRUMENT WITH DATA LOGGER, SERIAL INTERFACE, FOR BATTERY OPERATION OR OPTI ONAL M AINS SUPPLY, SINGLE INSTRUMENT 1 * 1 items
Catalog # 662-0502
Supplier:  WTW
PH/ION METER 340I,ROBUST AND WATERPROOF PH/ION HANDHELD INSTRUMENT WITH DATA LOGGER, SERIAL INTERFACE, FOR BATTERY OPERATION OR OPTI ONAL M AINS SUPPLY, SINGLE INSTRUMENT 1 * 1 items
Catalog # 662-0502
Supplier:  WTW
Frequently Bought Together