Orders LinkContactUsLinkComponent
Pipette tips
Pipette tips
  SARS70.3030.100
Marketsource
 :   SARSTEDT
Pipette tips
  SARS70.3030.100
 :  SARSTEDT
Marketsource
Pipette tips
67,42 €
1 * 1.920 ST
These high quality tips are suitable for the most common pipettes.
  • Sterile tips are DNase/RNase- and ATP-free, and non pyrogenic
  • Available loose in bags, packed in racks or in Tip StackPack reload systems
Tip StackPack: Tips are preloaded in reusable, stacked 96-tip trays. The trays are designed to safely cover each individual 96-tip layer, shielding the preloaded tips in the stack like a protective screen.
Accessories
  •  
    Description
    Colour
  • SARS95.1298.001
    Tip system box for pipette tip trays, with hinged lid, empty
    Transparent
    1 * 1 ST