Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Capillary columns, DB-624

on 5" cage, Capillary, DB-624, 3,00 µm, 0,53 mm, 30 m
Catalog # HEWL125-1334E
Supplier:   Agilent
Capillary columns, DB-624
Catalog # HEWL125-1334E
Supplier:  Agilent
Capillary columns, DB-624
Capillary columns, DB-624

on 5" cage, Capillary, DB-624, 3,00 µm, 0,53 mm, 30 m

Price
1 210,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These columns are recommended for EPA methods 502.2 and 8021, as well as for fast GC/MS volatiles analysis. They are specifically designed for the analysis of volatile priority pollutants and residual solvents.

  • Excellent inertness for active compounds
  • Bonded and cross-linked
  • Solvent rinsable
Specifications
  • Column Format:
    Capillary
  • Film thickness:
    3,00 µm
  • Ø int.:
    0,53 mm
  • Description:
    on 5" cage
  • Length:
    30 m
  • Phase:
    DB-624
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.