Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), 10 L
Catalog # BMOL54666-3
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Catalog # BMOL54666-3
CAS Number:  
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), 10 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 10 L
Product Details & Documents
Specifications

Specifications

  • Pk:
    10 L
  • Description:
    TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.