Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), Glass bottle, 1 L
Catalog # BMOL54666-1
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Catalog # BMOL54666-1
CAS Number:  
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), Glass bottle, 1 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)

Specifications