Order Entry
Belgium
Orders LinkContactUsLinkComponent
Agarose

Agarose

Glass bottle for solids, 10 g
Catalog # BMOL54540-1
Agarose
Catalog # BMOL54540-1
CAS Number:  9012-36-6
Agarose
Agarose

Glass bottle for solids, 10 g

CAS Number:  9012-36-6
Price
Inquire For Price
Log in for availability
1 * 10 g
Product Details & Documents
  • CAS Number:9012-36-6
  • MDL Number:MFCD00081294
  • Melting Pt:60…90 °C
  • Storage Temperature:Ambient
  • EINECS:232-731-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications