Hydroxyectoine
Glass bottle for solids, 1 g
Catalog # BMOL53239-2
Supplier: BIOMOL RESEARCH LABORATORIES
Jump to:
Product Details & Documents
- CAS Number:165542-15-4
- Formula:C₆H₁₀N₂O₃
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



