Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Packaging tapes

Fabric adhesive tape, Silver, 50,0 mm
  819-1411
 :   3M
Packaging tapes
  819-1411
 :  3M
Packaging tapes
Packaging tapes

Fabric adhesive tape, Silver, 50,0 mm

 
41,10 €
   
1 * 1 ROU
 

Universal adhesive tape for packaging, securing and labelling.

  • Excellent humidity resistance
  • Easy to tear during use
  • PE coating

Length: 50,0 m

 
  • Width:
    50,0 mm
  • Colour:
    Silver
  • Description:
    Fabric adhesive tape
  • Seq:
    02