Labels for slides
Slide labels on sheets, custom imprint, tag, Paper, Black on white, 22×22 mm
Catalogus # 631-9498
Leverancier: PRECISION DYNAMICS TIMEM
Ga naar:
Product Details & Documents
These thin labels are suitable for use with microscopy slides.
- Permanent adhesive
- No curling or detaching
Some models can be customised: Imprint can be on the top or bottom of the label
Specificaties
- Colour:Black on white
- Description:Slide labels on sheets, custom imprint, tag
- Dimensions:22×22 mm
- Material:Paper
- Seq:005