Cavity slides
Cavity slides with PTFE coating, White, 12, 5 mm
Catalog # 631-9423
Supplier: Epredia
Aller à :
Product Details & Documents
These autoclavable slides are specially designed for immunofluorescence.
- Adhesive surface and hydrophobic mask
- Hydrophobic epoxy- or PTFE-coating
- (*) with rings or wells
Developped for clinical diagnosis applications.
: Available with rings or wells (*).
Spécifications
- No. of wells:12
- Colour:White
- Description:Cavity slides with PTFE coating
- Seq:022
- Well Ø:5 mm